Precision thermometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L33012-TOL/VC L27378-FL/WC L27358-FL/WC L27378-FL L27334-FL/DKD3 L27304-FL L27306-FL L27308-GA L27308-GA/VC L27334-FL L27334-FL/VC L27336-FL L27354-CB L27358-FL L27374-CB L27302-FL L27302-FL/VC L27358-FL/DKD3
AMARL33012-TOL/VCEA 402 EUR
AMARL33012-TOL/VC AMARL27378-FL/WC AMARL27358-FL/WC AMARL27378-FL AMARL27334-FL/DKD3 AMARL27304-FL AMARL27306-FL AMARL27308-GA AMARL27308-GA/VC AMARL27334-FL AMARL27334-FL/VC AMARL27336-FL AMARL27354-CB AMARL27358-FL AMARL27374-CB AMARL27302-FL AMARL27302-FL/VC AMARL27358-FL/DKD3
Precision thermometers
Thermometers Analoge thermometers
These thermometers are suitable for government verification or calibration.
  • With ring at top
  • Durable pigment
  • Solid stem
Verpakking: Packed in a transparent square plastic case.