Body showers with eye/face shower, modular, wall-mounted
Leverancier: BROEN-LAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17552009 17555009 17535009
118-0552EA 4940 EUR
118-0552 BROE17555009 BROE17535009
Body showers with eye/face shower, modular, wall-mounted
Eerste-hulpbenodigdheden Douches
Eye shower, wall mounted in brass with chemical resistant BROEN-LAB Polycoat.
  • Professional solutions for employee safety
  • Easy to mount
  • Anti-limescale design and material
  • Improved functionality with long lifetime
  • Available with or without bowl
With built-in FLOWFIX for regulation of water flow (14 L/min) and stainless steel bowl for eye wash.
Min. working pressure: 1,5 bar.
Certificaten: EN 15154 (Part 1, 2)