Transfection reagent for viral capture, concentration and storage, Mag4C-AD
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ATK11200 AKC11000
OZBIATK11200EA 177.86 EUR
OZBIATK11200 OZBIAKC11000
Transfection reagent for viral capture, concentration and storage, Mag4C-AD
Transfectiereagentia
Mag4C-AD is designed for magnetic virus concentration. It has been specifically developed for capturing, concentrating and storing adenoviruses.
  • Simple, rapid and ready to use
  • Concentration of viruses by magnetic capture in 30 to 45 minutes
  • High yield of viral capture and recovery
  • No need to process magnetic beads before capture
  • Avoid ultracentrifugation, precipitation and chemicals, reduced handling steps of viruses
  • Improve virus stability under storage conditions
  • Ideal for cell culture infection, with all the advantages of Magnetofection™ technology
Mag4C magnetic nanoparticles capture viruses in culture media with 80-99% efficiency. Once captured onto magnetic beads, viruses can be:
Concentrated and stored with the storage buffer or directly used for downstream assay
Concentrated, eluted from the magnetic beads with the elution buffer and stored with the storage buffer or used for subsequent assays
The conservation buffers in the kit have been expressly designed to improve the stability of viruses under storage conditions and are fully compatible with the magnetic nanoparticles.
Informatie over toebehoren: Multipurpose magnetic separation rack for 50, 15 or 1,5 ml tubes needs to be used with the kit (available separately).
Opgelet: Store at +4 °C.