Luciferase assay kit
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LUC0100 LUC1000
OZBILUC0100EA 126.85 EUR
OZBILUC0100 OZBILUC1000
Luciferase assay kit
Testen Cellulaire assays
The luciferase assay kit is the most sensitive analytical tools for measuring gene expression.
  • Very sensitive and quantitative assay with a wide dynamic range
  • Bioluminescence measurements are performed directly into transfected wells
  • Cell lysis buffer is optimised for Luciferase assay buffer to ensure maximal sensitivity
  • Light emission is proportional to number of luciferase enzyme molecule
Kit contains optimised components (cell lysis buffer, luciferase substrate and luciferase assay buffer) to ensure maximal sensitivity with a consistent light output.
Fast and easy, accurate and linear, this kit detects and quantify firefly luciferase in transfected eukaryotic cells reaching high sensitivity level. Since the light emission is constant (>15 min) when using this kit, there is no need to use an automated luminometer for substrate injection. Emitted light is recorded by a standard luminometer at 560 nm.