Web self-retracting lifeline, DBI-SALA® Nano-Lok™, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3101208 3101298
115-2675EA 0 EUR
115-2675 115-2676
Web self-retracting lifeline, DBI-SALA® Nano-Lok™, 3M™
Valbescherming Veiligheidskoorden en -touwen
Nano-Lok™ is compact and ergonomically designed for ease-of-use, and is ideal as a direct connection to most harnesses. Weighing only 725g, it is virtually unnoticeable when worn. It can be used as a lanyard replacement. Locks quickly and stops a fall within centimetres, providing more protection at low heights.
  • Single leg harness connector
  • Swiveling anchorage loop
  • Impact-resistant housing
  • Compatible with a shoulder or foot connection
  • Energy absorber/impact indicator
  • 2m retractable lifeline
  • Aluminum twin connector and carabiner of 57mm
  • 100% tie-off configuration
Compatible with shoulder and foot level connections and available in a number of configurations, with a capacity of 141kg, Nano-Lok™ offers versatility for many applications. In addition, tension is always kept on the lifeline to reduce dragging, snagging, and trip falls. Ideal for direct connection to most harnesses.
Gate (1630kg) reduces chances of accidental disengagement (roll-out) for added safety.
Certificaten: EN 360