Aluminium tripod, DBI-SALA, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8563158 8563159
CPSD8563158EA 0 EUR
CPSD8563158 CPSD8563159
Aluminium tripod, DBI-SALA, 3M™
Valbescherming Veiligheidslijnen
Tripod with adjustable locking legs, safety chains, top pulley and winch bracket.
  • Available in two heights
Weight capacity: 205 kg
Certificaten: EN 795 Class-B
Bestelinformatie: Winch and SRL are not included.