Retrieval self-retracting lifeline, Sealed-Block™ DBI-SALA, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3400856 3400881
CPSD3400856EA 0 EUR
CPSD3400856 CPSD3400881
Retrieval self-retracting lifeline, Sealed-Block™ DBI-SALA, 3M™
Valbescherming Veiligheidslijnen
Sealed-Blok™ family has the most comprehensive line of sealed SRLs in the industry and decades of proven field service in the toughest environments.
  • Pivoting anchorage loop
  • Aluminum housing and stainless steel life line
  • 2267 kg Rated handle
  • Impact indicator
  • Tripod / davit arm bracket option
  • 5 mm Ø rope and 1633 kg gate
  • RSQ options and FAST-Line lifeline assembly
The heavy duty sealed design keeps the critical working components free of dirt, grease, water and chemicals. The range also includes a dual range SRL and automatic self-rescue device. Ideal for use in harsh environments such as offshore, confined space, salt water and dirty environments.
Certificaten: Meets IP68 rating, EN 360, EN1496 clB.