Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride, RuClCp(PPh3)
46430.03 46430.06
46430.03EA 184 EUR
46430.03 46430.06
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.
Moisture Sensitive. Air Sensitive. Store under Argon.
Formule: C₄₃H₄₁P₂ClORu
MW: 772.27 g/mol
Smeltpunt: 135 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075004
CAS nummer: 32993-05-8