ESD occupational shoes, lace-up, Berlin
Leverancier: SCHURR SCHUHVERTRIEB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11640-02-9436 11640-02-9437 11640-02-9438 11640-02-9439 11640-02-9440 11640-02-9441 11640-02-9442 11640-02-9443 11640-02-9444 11640-02-9445 11640-02-9446 11640-02-9447 11640-02-9448
SCRR11640-02-9436EA 103 EUR
SCRR11640-02-9436 SCRR11640-02-9437 SCRR11640-02-9438 SCRR11640-02-9439 SCRR11640-02-9440 SCRR11640-02-9441 SCRR11640-02-9442 SCRR11640-02-9443 SCRR11640-02-9444 SCRR11640-02-9445 SCRR11640-02-9446 SCRR11640-02-9447 SCRR11640-02-9448
ESD occupational shoes, lace-up, Berlin
Schoenen en laarzen
Occupational shoe made from leather/polyester upper, an antibacterial lining and a slip resistant and profiled nitrile/rubber EXPERT sole.
  • POROMAX®3D-Mesh, antibacterial lining
  • PUR-foam with DRYtex plus fabric, antibacterial, fungicide, ESD insole
  • Replacable and orthopaedic insoles possible
  • Triple cushioning plus: Shock absorbing Technogel® pads in the forefoot and heel area
Suitable for use in kitchens and in the food industry.
Colour: Black; weight: 420 g; width: 10
Certificaten: DGUV 112-191; EN ISO 20347:2012; O1, SRC