Sterilisation gusseted reels, Stericlin®
Leverancier: VP Medical Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3FKSS230216 3FKSS230218 3FKSS230212 3FKSS230204 3FKSS230210 3FKSS230206 3FKSS230214
129-1287EA 232 EUR
129-1287 129-1288 129-1285 129-1282 129-1284 129-1283 129-1286
Sterilisation gusseted reels, Stericlin®
Zakken Sterilisatiezakken
These see-through sterilisation reels made of sterilisation proof 70 g/m2 heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certificaten: DIN EN ISO 11607 and DIN 868-5.