HPLC columns, NUCLEODUR® NH2 / NH2-RP
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
760732.46 760730.46 760732.40 761953.30 760732.30 760742.46 760722.46 760741.20 760730.30 761952.30 760722.40 760720.40 760720.46 760721.46 760730.20 760730.40 760731.46 760732.20 760739.46 760740.20 761951.20 761951.30 761953.20
MANA760732.46EA 511 EUR
MANA760732.46 MANA760730.46 MANA760732.40 MANA761953.30 MANA760732.30 MANA760742.46 MANA760722.46 MANA760741.20 MANA760730.30 MANA761952.30 MANA760722.40 MANA760720.40 MANA760720.46 MANA760721.46 MANA760730.20 MANA760730.40 MANA760731.46 MANA760732.20 MANA760739.46 MANA760740.20 MANA761951.20 MANA761951.30 MANA761953.20
HPLC columns, NUCLEODUR® NH2 / NH2-RP
Kolommen voor chromatografie
NUCLEODUR® NH2 and NH2-RP are aminopropyl modified silica phase for HPLC.
  • High purity spherical silica gel
  • Weakly hydrophobic, polar and ionic phase with applications for NP, RP and IC separations (multi-mode)
  • Separation mechanism based on polar, hydrophobic and weak ion exchange interactions
Polar analytes like sugars or DNA bases can be separated in reversed phase mode (RP) with aqueous-organic eluent systems. NUCLEODUR® NH2-RP columns containing water – acetonitrile as eluent are ready-to-use without a reflushing. Polar compounds like substituted anilines, esters, and chlorinated pesticides will be analysed in normal phase mode (NP) with n-heptane, dichloromethane or 2-propanol as mobile phase. The suitable NUCLEODUR® NH2 columns contain n-heptane eluent. For ion chromatography (IC) of anions and organic acids the aminopropyl phase is applied with conventional buffers and organic modifiers.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).