Petroleum standards, polywax
Leverancier: Restek
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36224
36225
36227
REST36224EA
39.5
EUR
REST36224
REST36225
REST36227
Petroleum standards, polywax
Standaarden
Standaarden voor de petroleumindustrie
These high molecular weight hydrocarbon waxes are useful for simulated distillation and other high temperature GC work.
Supports ASTM Methods D2887, D6352, D7169, D7398, and D7500.