Petroleum standards, polywax
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36224 36225 36227
REST36224EA 39.5 EUR
REST36224 REST36225 REST36227
Petroleum standards, polywax
Standaarden Standaarden voor de petroleumindustrie
These high molecular weight hydrocarbon waxes are useful for simulated distillation and other high temperature GC work.
Supports ASTM Methods D2887, D6352, D7169, D7398, and D7500.