Stainless steel, wire gauzes with frame for gas burners
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
247.935 247.955 247.995 247.975
RSGA247.935EA 47.8 EUR
RSGA247.935 RSGA247.955 RSGA247.995 RSGA247.975
Stainless steel, wire gauzes with frame for gas burners
Stainless steel
Wire mesh with frame
  • Stainless steel 1.4301
  • mesh: 0.6 x 2.0 mm
  • fine mesh centre: Heat resistant stainless steel