Safety needles, Sterican®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4670005S-01 4670035S-01 4670012S-01 4670016S-01 4670025S-01 4670002S-01 4670008S-01 4670045S-01 4670050S-01 4670052S-01 4670055S-01
BRAU4670005S-01EA 64 EUR
BRAU4670005S-01 BRAU4670035S-01 BRAU4670012S-01 BRAU4670016S-01 BRAU4670025S-01 BRAU4670002S-01 BRAU4670008S-01 BRAU4670045S-01 BRAU4670050S-01 BRAU4670052S-01 BRAU4670055S-01
Safety needles, Sterican®
Injectienaalden
This single-use needle with safety protection is compatible with all standard Luer lock and Luer slip syringes.
  • Made of stainless chromium-nickel steel
  • Needle hub made of PP
The safety feature of Sterican® safety needle is in compliance with the EU Directive and provides an intuitive, one-handed activation. It securely and irreversibly locks the needle within the integrated shield for a safe disposal.
Certificaten: In compliance with EU Directive 2010/32/EU. Colour coding according to ISO 6009.