Bulk media for HPLC, NUCLEODUR® C18 HTec
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
713831.0100 713832.0100
MANA713831.0100EA 5140 EUR
MANA713831.0100 MANA713832.0100
Bulk media for HPLC, NUCLEODUR® C18 HTec
Sorbenten voor chromatografie Sorbenten voor HPLC
High purity silica phase for preparative HPLC.
  • Totally spherical high purity silica
  • Pressure stable up to 600 bar
  • Pore volume 0,9 ml/g
  • Surface (BET) 340 m²/g
  • Density 0,47 g/ml
  • Carbon content 18%
  • Reliable and durable RP phase for up-scaling to preparative scale
The high density monomeric octadecyl silica NUCLEODUR® C18 HTec is recommended for the preparative separartion of basic, neutral and acidic substances (pharmaceuticals, pesticides, vitamins).