ASTM precision thermometers, with DAkkS calibration certificate
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1202089S/04/3P 1202090S/04/3P 1202092S/04/3P 1202093S/04/3P 1202094S/04/3P 1202095S/04/3P 1202096S/04/3P 1202054S/04/3P 1202012S/03/3P 1202092S/03/3P 1202033S/03/3P 1202071S/04/4P 1202014S/04/5P 1202046S/04/3P 1202054S/04/4P 1202061S/04/5P 1202121S/04/3P 1202061S/04/3P 1202005S/03/3P 1202015S/03/3P 1202018S/03/3P 1202022S/03/3P 1202056S/03/3P 1202058S/03/3P 1202059S/03/3P 1202062S/03/3P 1202063S/03/3P 1202064S/03/3P 1202065S/03/3P 1202066S/03/3P 1202067S/03/3P 1202091S/03/3P 1202116S/03/3P 1202117S/03/3P 1202120S/03/3P 1202130S/03/3P 1202022S/04/3P 1202056S/04/3P 1202058S/04/3P 1202059S/04/3P 1202063S/04/3P 1202064S/04/3P 1202065S/04/3P 1202066S/04/3P 1202067S/04/3P 1202116S/04/3P 1202117S/04/3P 1202130S/04/3P
SCNL1202089S/04/3PEA 572 EUR
SCNL1202089S/04/3P SCNL1202090S/04/3P SCNL1202092S/04/3P SCNL1202093S/04/3P SCNL1202094S/04/3P SCNL1202095S/04/3P SCNL1202096S/04/3P SCNL1202054S/04/3P SCNL1202012S/03/3P SCNL1202092S/03/3P SCNL1202033S/03/3P SCNL1202071S/04/4P SCNL1202014S/04/5P SCNL1202046S/04/3P SCNL1202054S/04/4P SCNL1202061S/04/5P SCNL1202121S/04/3P SCNL1202061S/04/3P SCNL1202005S/03/3P SCNL1202015S/03/3P SCNL1202018S/03/3P SCNL1202022S/03/3P SCNL1202056S/03/3P SCNL1202058S/03/3P SCNL1202059S/03/3P SCNL1202062S/03/3P SCNL1202063S/03/3P SCNL1202064S/03/3P SCNL1202065S/03/3P SCNL1202066S/03/3P SCNL1202067S/03/3P SCNL1202091S/03/3P SCNL1202116S/03/3P SCNL1202117S/03/3P SCNL1202120S/03/3P SCNL1202130S/03/3P SCNL1202022S/04/3P SCNL1202056S/04/3P SCNL1202058S/04/3P SCNL1202059S/04/3P SCNL1202063S/04/3P SCNL1202064S/04/3P SCNL1202065S/04/3P SCNL1202066S/04/3P SCNL1202067S/04/3P SCNL1202116S/04/3P SCNL1202117S/04/3P SCNL1202130S/04/3P
ASTM precision thermometers, with DAkkS calibration certificate
Thermometers Analoge thermometers
These precision thermometers are ideal to measure precisely and reliably without mercury from the very start.
  • Round, white back capillary form
  • Suitable for calibration
Blue filling stem type.
Certificaten: Conform to ASTM standards.
Bestelinformatie: Available with works certificate or with DAkkS calibration certificate (optional).
Verpakking: Packed individually in a square plastic sleeve.