Trident LC column protection system
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27472 27474 27475
REST27472EA 164 EUR
REST27472 REST27474 REST27475
Trident LC column protection system
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This column protection system has three levels of protection: filter only, cartridge only, or filter and cartridge.
  • Durable metal tip with replaceable PEEK ferrule means easy installation onto column without tools
  • Improved thread design and materials create an optimal seal that releases and reseals easily
  • Allows multiple installations without galling and binding
  • Easy-to-remove cap frit simplifies filter replacement
  • Low-dead-volume design has negligible effect on chromatography
  • Direct connection eliminates tubing and connectors that increase system volume