Fuel oil #6 standards
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31218 31248 31249
REST31218EA 51 EUR
REST31218 REST31248 REST31249
Fuel oil #6 standards
Standaarden Standaarden voor de petroleumindustrie
A high-viscosity residual type of heavy fuel oil requiring preheating to 104 to 127 °C. This material is also known as residual fuel oil (RFO) or Bunker C.
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.