Jumbo syringes
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21275 21276 21277
REST21275EA 1330 EUR
REST21275 REST21276 REST21277
Jumbo syringes
Spuiten Injectiespuiten voor chromatografie
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
  • Central port with a Luer lock fitting
  • Secondary port with septum nut
  • Detachable plunger stem
  • Easy sample storage
An adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time.