Gas-tight syringe replacement needles for removable needle syringes
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24803 24804 24805 24806 24807 24802
REST24803EA 0 EUR
REST24803 REST24804 REST24805 REST24806 REST24807 REST24802
Gas-tight syringe replacement needles for removable needle syringes
Injectienaalden
For use with removable needle syringes.
  • Stainless steel needle
  • Nickel plated brass hub