Perchloorzuur 0.1 M (0.1 N) in glacial acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
35418-1L 35418-500ML
35418-1LEA 158 EUR
35418-1L 35418-500ML
Perchloorzuur 0.1 M (0.1 N) in glacial acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Perchloorzuur
Formule: HClO₄
MW: 100.46 g/mol
Kookpunt: 39
Smeltpunt: -101
Dichtheid: 1.06 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011325
CAS nummer: 7601-90-3
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 0.0997 - 0.1003 mol/l
Water (Karl Fischer) 0.1 - 0.2 %
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective