Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
270733-1L 270733-2.5L 270733-4L 270733-6X1L
270733-1LEA 71.2 EUR
270733-1L 270733-2.5L 270733-4L 270733-6X1L
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Water
  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Formule: H₂O
Kookpunt: 100 °C
Smeltpunt: 0 °C
Dichtheid: 0,9971 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now
Specification Test Results
Non-volatile matter Max. 0.0003 %
Absorbance at 400 nm Max. 0.005
Absorbance at 250 nm Max. 0.005
Absorbance at 210 nm Max. 0.01
Absorbance at 205 nm Max. 0.01
HPLC Gradient at 254 nm Max. 2 mAU
HPLC Gradient at 210 nm Max. 10 mAU