Natriumhydroxide 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Caustic soda
35263-1L 35263-5L-VP 35263-10L-VP 35263-5L 35263-6X1L
35263-1LEA 31 EUR
35263-1L 35263-5L-VP 35263-10L-VP 35263-5L 35263-6X1L
Natriumhydroxide 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Natriumhydroxide
Formule: NaOH
Kookpunt: 100 °C (1,013 hPa)
Dichtheid: 1,000 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective