Natriumhydroxide concentrate for dilution to 1 L, 1.0 M NaOH (1.0 N) Volumetric, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Caustic soda
38215-1EA 38215-6X1EA
38215-1EAEA 50.9 EUR
38215-1EA 38215-6X1EA
Natriumhydroxide concentrate for dilution to 1 L, 1.0 M NaOH (1.0 N) Volumetric, Fluka™
Natriumhydroxide
Concentrate for 1 L standard solution
Formule: NaOH
MW: 40 g/mol
Kookpunt: 120 °C
Smeltpunt: 323
Dichtheid: 1.350 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Specification Test Results
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective