Natriumthiosufaat concentrate for dilution to 1L, 0.01 M Na2S2O3 (0.01N) volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
38243-1EA 38243-6X1EA
38243-1EAEA 58.5 EUR
38243-1EA 38243-6X1EA
Natriumthiosufaat concentrate for dilution to 1L, 0.01 M Na2S2O3 (0.01N) volumetric solution, Fluka™
Natriumthiosufaat
Concentrate for 1L standard solution
Formule: Na₂S₂O₃
MW: 158.11 g/mol
Kookpunt: 100 °C (1.013 hPa)
Smeltpunt: 48
Dichtheid: 1.0 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
Merck Index: 12,08844
Order Now
Specification Test Results
Factor 0.998 - 1.002
Specificaties
Environmentally Preferable