Dithalliumsulfaat ≥98.0% (RT), Purum, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

88290-100G 88290-500G
88290-100GEA 235 EUR
88290-100G 88290-500G
Dithalliumsulfaat ≥98.0% (RT), Purum, Fluka™
Dithalliumsulfaat

  • ≥95% for general laboratory use.

Formule: O₄STl₂
Kookpunt: Dec. (on heating)
Smeltpunt: 632 °C
Dichtheid: 6,77 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011278
CAS nummer: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II
Merck Index: 12,09405

Order Now

Specification Test Results

Assay (bromatometric) 98.0 - 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR