N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

33120-180KG 33120-1L 33120-2.5L 33120-25L 33120-4X2.5L 33120-6X1L 33120-2.5L-PE 33120-7L-RC
33120-180KGEA 2840 EUR
33120-180KG 33120-1L 33120-2.5L 33120-25L 33120-4X2.5L 33120-6X1L 33120-2.5L-PE HONC33120-7L-RC
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, Riedel-de Haën™
N,N-Dimethylformamide

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 605365
RTECS Number: LQ2100000
UNSPSC Code: 12191602

Formule: HCON(CH₃)₂
Kookpunt: 152-153 °C (1,013 hPa)
Smeltpunt: -61 °C
Dichtheid: 0,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Density (D 20/4) 0.948 - 0.950
Refractive index (n 20/D) 1.4290 - 1.4310
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Titrable Base Max. 0.003 meq/g
Titrable Acid Max. 0.0005 meq/g
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.0001 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
APHA Max. 15
Refractive Index n20/D 1.4290-1.4310


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR