Zoutzuur 1 M HCl (1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrochloric acid solution, Muriatic acid
35328-1L 35328-5L 35328-10L-VP 35328-30L 35328-4X5L 35328-6X1L 35328-5L-VP
35328-1LEA 30.7 EUR
35328-1L 35328-5L 35328-10L-VP 35328-30L 35328-4X5L 35328-6X1L 35328-5L-VP
Zoutzuur 1 M HCl (1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Zoutzuur
Formule: HCl
MW: 36.46 g/mol
Kookpunt: 100 °C (1.013 hPa)
Smeltpunt: 0 °C
Dichtheid: 1.010 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-791-2
UN: 1789
ADR: 8 III
Order Now
Specification Test Results
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective