Mineral spirits standards (unweathered)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31225 31260
REST31225EA 50 EUR
REST31225 REST31260
Mineral spirits standards (unweathered)
Standaarden Standaarden voor de petroleumindustrie
This standard is manufactured using the Type 1 mineral spirit.
There are four general types of mineral spirits, classified according to boiling point range (BPR):
Type I (Stoddard solvent) BPR 149 to 182 °C
Type II (high fl ash point) BPR 177 to 196 °C
Type III (odorless) BPR 149 to 196 °C
Type IV (low dry point) BPR 149 to 174 °C.
Certificaten: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
Specificaties
Environmentally Preferable