Pharmacist's spatulas
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3500 3501 3502 3503 3504
BOCH3500EA 6.8 EUR
BOCH3500 BOCH3501 BOCH3502 BOCH3503 BOCH3504
Pharmacist's spatulas
Spatels
These spatulas feature a stainless steel blade and a PVC handle.
  • Suitable for dishwasher
  • Autoclavable