Bottle-top dispensers, DISPENSMAN™
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F110101 F110102 F110103 F110104 F110105
GILSF110101EA 385 EUR
GILSF110101 GILSF110102 GILSF110103 GILSF110104 GILSF110105
Bottle-top dispensers, DISPENSMAN™
Flesdispensers
Variable volume, analogue-adjustable.
  • Quick, easy, and precise volume setting
  • Recycle position avoids any loss of liquid
  • Recirculation valve for purging air
  • Anti-drip system for keeping the lab bench dry
  • Simple operation with only one hand
  • Do not use a motor or require electricity: Flexible usage
These bottle-top dispensers offer precise, quick and safe liquid dispense directly from the bottle to the final receptacle.
A variety of bottle adapters are provided with DISPENSMAN to connect to various bottles to dispense almost any liquid for manyMore Product Information