Labels for BradyJet J2000 SFID printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
150008 150012 150030 150003 150009 150010 150013 150014 150017 150021 150026 150027 150029 150031 150032
BRDY150008EA 191.6 EUR
BRDY150008 BRDY150012 BRDY150030 BRDY150003 BRDY150009 BRDY150010 BRDY150013 BRDY150014 BRDY150017 BRDY150021 BRDY150026 BRDY150027 BRDY150029 BRDY150031 BRDY150032
Labels for BradyJet J2000 SFID printer
Etiketten
These BradyJet J2000 SFID printer labels are used to produce eye-catching safety signs, tags, pipe markers and lean instruction labels to increase workplace safety and efficiency and at the same time comply with any regulatory colour code.
  • Stay attached and remain legible when exposed to liquids, alcohol and indoor lighting
  • Different sizes and materials available
They can be counted on to warn coworkers about hazards, or to instruct them on the optimal use or maintenance of machinery.