Diesel fuel #2 composite standard
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31258 31259
REST31258EA 79.5 EUR
REST31258 REST31259
Diesel fuel #2 composite standard
Standaarden Standaarden voor de petroleumindustrie
Diesel fuel composite standards are comprised of three separate sources blended in equal portions. It may exhibit lot-to-lot variation.
Certificaten: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements