Stand rods
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2000/2100 2002/2102 2003/2103 2004/2104
USBE2000/2100EA 31 EUR
USBE2000/2100 USBE2002/2102 USBE2003/2103 USBE2004/2104
Stand rods
Statiefstaven
Steel, zinc plated.
  • Threaded (M 10)
  • Diameter: 12 mm