UltraSpeed microLite mop
Leverancier: Vileda Professional
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VILE143210EA 14.4 EUR
VILE143210
UltraSpeed microLite mop
Moppen
Ultra Speed is the complete wet flat mopping system that combines intensive microfibre cleaning with a revolutionary high performance press. The system is so easy to use that anyone can start cleaning without the need for training. The unique press wrings out more water than traditional gear presses, allowing floors to dry faster and be back in use quicker - vital in high traffic areas.
  • Low overall weight
  • MicroLite mop can be machine washed over 250 times at 70 °C
  • Reduces laundry costs
An effective, easy to use all-purpose microfibre mop for general cleaning of sealed and smooth floors. The combination of 100% polyester microfibre material and polyamide abrasive strips easily removes stubborn dirt.