Filtration cartridges, CHROMAFIL®
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
730517.3100 730517.6100 730570.320 730570.345 730570.620 730570.645 730578.620 730578.645
MANA730517.3100EA 185 EUR
MANA730517.3100 MANA730517.6100 MANA730570.320 MANA730570.345 MANA730570.620 MANA730570.645 MANA730578.620 MANA730578.645
Filtration cartridges, CHROMAFIL®
Filters Filterpatronen
These filtration cartridges are used for sample clarification under vacuum (e.g., using the CHROMABOND® vacuum manifold or SPE automation systems) or by gravity. By using CHROMAFIL® filtration cartridges with their specific membranes unwanted particles are removed which can otherwise cause contamination and clogging of sensitive instrumentation.
  • Cartridge sizes 3 and 6 ml
  • Available with different membranes (PET, RC, PTFE, PVDF, GF) and pore sizes (0,20; 0,45 and 1,00 µm)
PET membrane: Hydrophilic multipurpose polyester membrane for polar as well as nonpolar solvents; especially suited for mixtures of water and organic solvents (HPLC eluents).
RC membrane: Hydrophilic regenerated cellulose membrane for aqueous and organic-aqueous liquids like polar and medium polar sample solutions.
PTFE membrane: Hydrophobic polytetrafluoroethylene membrane for nonpolar liquids and gases; very resistant towards all kinds of solvents as well as acids and bases.
PVDF membrane: hydrophilic polyvinylidene difluoride membrane for 100% aqueous samples; water-soluble oligomers and polymers like proteins.
GF membrane: inert glass fiber filter with nominal pore size of 1 µm allows higher flow rates than small pore filters; for solutions with high loads of particulate matter or for highly viscous solutions like soil samples, and fermentation broth.