Gloves, natural rubber, Sicco
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V1972-07 V1972-08 V1972-09
BOHLV1972-07EA 540 EUR
BOHLV1972-07 BOHLV1972-08 BOHLV1972-09
Gloves, natural rubber, Sicco
Handschoenen
These natural rubber gloves are wearable on both sides.
  • Perforation and abration proof
  • High flexibility
  • Low voltage value
  • Low electrostatic charging