ESD occupational shoes, lace-up, Uni6, 36761
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36761/36 36761/39 36761/40 36761/37 36761/38 36761/45 36761/46 36761/47 36761/48 36761/43 36761/44 36761/41 36761/42 36761/35
ABEB36761/36EA 113 EUR
ABEB36761/36 ABEB36761/39 ABEB36761/40 ABEB36761/37 ABEB36761/38 ABEB36761/45 ABEB36761/46 ABEB36761/47 ABEB36761/48 ABEB36761/43 ABEB36761/44 ABEB36761/41 ABEB36761/42 ABEB36761/35
ESD occupational shoes, lace-up, Uni6, 36761
Schoenen en laarzen
ESD shoes made from functional leather with honeycomb pattern and a TPU sole resistant to oils, fuels and animal fats.
  • Breathable lining with silver ions
  • Replaceable Soft Comfort insole M; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • PU midsole
  • Slip resistant TPU sole
  • Water repellent and scratch proof
Colour: Black
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O2, FO, SRC