Sample boxes with snap-on lid, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EP28.1 EP22.1 EP24.1 EP26.1
ROTHEP28.1EA 158 EUR
ROTHEP28.1 ROTHEP22.1 ROTHEP24.1 ROTHEP26.1
Sample boxes with snap-on lid, Rotilabo®
Monsternamepotjes Containers voor algemeen gebruik
Rotilabo® sample boxes, made of PP and with attached lid.
  • With labelling area
  • Graduated by 10 ml steps (except Cat. No. ROTHEP22.1)
  • Leakproof
  • Autoclavable with open lid
Temperature stable from –20 to +121 °C.