Teclu safety burner
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VL01374DE VL01374DP VL01374P
JUCHVL01374DEEA 0 EUR
JUCHVL01374DE JUCHVL01374DP JUCHVL01374P
Teclu safety burner
Verwarmers Gasbranders
Teclu safety burner with needle valve and thermofeeler with safety valve on heavy, round base.
Height: 200 mm
Weight: 685 g