DURAN® PURE GLS 80® Bottles
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
818665405 818666307 818604403 818605408 818606301 818664409 818609101 818607306 818608602 818667303 818668608 818669107 818666907
SCOT818665405EA 939 EUR
SCOT818665405 SCOT818666307 SCOT818604403 SCOT818605408 SCOT818606301 SCOT818664409 SCOT818609101 SCOT818607306 SCOT818608602 SCOT818667303 SCOT818668608 SCOT818669107 SCOT818666907
DURAN® PURE GLS 80® Bottles
Flessen Glazen flessen
DURAN® PURE bottles clear or amber, with GLS 80 thread and dust cover. DURAN PURE is a range of primary packaging products dedicated to the specific requirements and needs of the pharmaceutical industry.
  • Made from DURAN® borosilicate glass 3.3 Type I
  • Wide mouth allows easy filling and emptying of bottles
Suitable for manufacturing of pharmaceutical products, storage of bulk drug intermediates and ingredients and transportation of API.
Certificaten: The bottles and closures in the range fully meet the requirements laid down in the international pharmacopoeias, and offer full protection for sensitive and valuable pharmaceutical products. DURAN PURE products are available with a full range of documents and certification that meet the detailed requirements of the regulatory authorities.