High Visibility Craftsman Trousers, Fristads® 2029 PLU
Leverancier: Fristads Kansas Group
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fristads®
KWIN100978271C60EA 84.6 EUR
KWIN100978271C60 KWIN100978271C62 KWIN100978271C64 KWIN100978271C156 KWIN100978271D108 KWIN100978271D104 KWIN100978271C44 KWIN100978271C66 KWIN100978271C46 KWIN100978271C48 KWIN100978271D92 KWIN100978271C50 KWIN100978271D96 KWIN100978271C52 KWIN100978271C150 KWIN100978271D120 KWIN100978271D100 KWIN100978271C154 KWIN100978271C152 KWIN100978271D116 KWIN100978271C146 KWIN100978271D112 KWIN100978271C148 KWIN100978271C54 KWIN100978271C56 KWIN100978271C58
High Visibility Craftsman Trousers, Fristads® 2029 PLU
Werkkleding
High visibility trousers in dirt, oil and water repellent material (80% polyester, 20% cotton) with soft cotton inside and CORDURA® reinforcements at exposed areas.
  • Two tuckable CORDURA® reinforced loose-hanging front pockets, left pocket with extra pocket and right pocket with three pockets and tool loops
  • Two front pockets and two back pockets (one with flap and velcro fastening)
  • D-ring at waistband, double reinforced crotch seam
  • Leg pocket with flap and velcro fastening, phone pocket with flap and velcro fastening, D-ring and plastic ID-card holder
  • Hammer loop; CORDURA® reinforced folding rule pocket with tool pocket, two buttons and loops for two sheath knifes
  • Pre-shaped knees, CORDURA® reinforced knee pockets with inside opening
  • Height adjustment for knee pads in knee pocket
  • CORDURA® reinforced leg ends
Weight: 300 g/m²
Certificaten: EN ISO 20471 (class 1)
OEKO-TEX® certified