Flame-retardant trousers, Flamestat 2144 ATHS
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121354540C50 121354540C52 121354540C44 121354540C54 121354540C60 121354540C62 121354540C56 121354540C58 121354540C46 121354540C48
KWIN121354540C50EA 229.9 EUR
KWIN121354540C50 KWIN121354540C52 KWIN121354540C44 KWIN121354540C54 KWIN121354540C60 KWIN121354540C62 KWIN121354540C56 KWIN121354540C58 KWIN121354540C46 KWIN121354540C48
Flame-retardant trousers, Flamestat 2144 ATHS
Werkkleding
Dirt, oil and water repellent trousers with inherent flame protection adapted for ATEX environments with no metal details. Manufactured from 45% modacrylic, 35% cotton, 18% polyamide and 2% anti-static fibre, reinforcements are made from 54% modacrylic, 44% cotton and 2% anti-static fibre. Approved for industrial laundry.
  • Two front pockets; D-ring at waistband
  • Double reinforced crotch seam
  • Two back pockets with flap
  • Leg pocket with flap and concealed snap fastening, phone pocket with flap and velcro fastening, D-ring and detachable plastic ID-card holder (holder contains metal)
  • Leg pocket with flap; Ruler pocket
  • Pre-shaped knees; CORDURA® reinforced knee pockets with inside opening; height adjustment for knee pads in knee pocket
  • Adjustable leg length with 5 cm hem allowance
Weight: 300 g/m²
Certificaten: Approved according to EN 14404 together with kneepads 124292, EN 61482-1-2 (class 1), EN 61482-1-1 EBT: 12,8 cal/cm² HAF: 84,2 %, EN ISO 11612 (A1 B1 C1 E2 F1), EN ISO 11611 A1 (class 1), EN 1149-5, EN 13034 Type PB [6]
OEKO-TEX® certified