Cooling plate, HistoCore arcadia C
Leverancier: Leica
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LEIC14039353599EA 0 EUR
LEIC14039353599
Cooling plate, HistoCore arcadia C
Koelers Tafelmodel koelers
HistoCore Arcadia C is a cold plate holding more than 60/65 cassettes on its large working surface. Cooling efficiency is important, so the cold plate was designed with an environment adaptive control module to make sure the operating temperature is always stabilized at –6 °C. There is no need to turn down the temperature in summer or worry about too fast cooling in winter.
  • Smooth workflow, simple operation and precise control
  • Wrist pads allow for increased comfort and stability
  • Easy to clean metal frame and silicon coated wristpads
  • The sturdy design of the special scraper helps clean all grooves and crevices
The Arcadia C cold plate can be used as a stand-alone unit for re-cooling blocks prior to sectioning or as part of an embedding center to cool molds after paraffin dispensing.