Overalls with bib and braces, Kansas® ICON One, 1111 KC
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113101530C56 113101530C58 113101530C60 113101530C62 113101530C44 113101530C46 113101530C48 113101530C64 113101530C66 113101530C52 113101530C54 113101530C50
KWIN113101530C56EA 95.9 EUR
KWIN113101530C56 KWIN113101530C58 KWIN113101530C60 KWIN113101530C62 KWIN113101530C44 KWIN113101530C46 KWIN113101530C48 KWIN113101530C64 KWIN113101530C66 KWIN113101530C52 KWIN113101530C54 KWIN113101530C50
Overalls with bib and braces, Kansas® ICON One, 1111 KC
Werkkleding
Bib'n'brace for industrial work in durable and comfortable 65% polyester/35% cotton material.
  • Three-needle fell seams, contrast stitching, diagonal design on back
  • Knitted braces, zip fly
  • Adjustable waist, adjustable leg length with 5 cm hem allowance
  • Side opening to inner trousers
  • Reflective detailing on back of knees
  • Bib pocket with flap, inside bib pocket with zip, two side pockets (right with integrated mobile phone pocket)
  • Large leg pocket with flap - placed over side seam - with mobile phone pocket with flap and pen pockets
  • Double folding rule pocket - placed over side seam - with pen pocket
  • Two back pockets, one with flap
  • Hammer loops, neck flap for barcode or chip
Weight: 295 g/m²