Specific gravity hydrometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H801850 H801852 H801854 H801856 H801858 H801860 H801862 H801864 H801866 H801868 H801870 H801872 H801874 H801876 H801890 H801892 H801894 H801896 H801898 H801900 H801902 H801920 H801922 H801924
AMARH801850EA 26.3 EUR
AMARH801850 AMARH801852 AMARH801854 AMARH801856 AMARH801858 AMARH801860 AMARH801862 AMARH801864 AMARH801866 AMARH801868 AMARH801870 AMARH801872 AMARH801874 AMARH801876 AMARH801890 AMARH801892 AMARH801894 AMARH801896 AMARH801898 AMARH801900 AMARH801902 AMARH801920 AMARH801922 AMARH801924
Specific gravity hydrometers
Hydrometers
These hydrometers are designed for the ranges of 0,600 up to 2,000 °SG.
  • Without thermometer
  • Paper scale
  • Shot weighted body
Reference temperature: 28,9/28,9 °C resp. 84 °F.
Leveringsinformatie: Supplied in a transparent plastic case.