Eyewash bottle, sterilised, diphoterine, Esculape
Leverancier: Laboratoires Esculape
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Diphoterine®
LBES8205020EA 68 EUR
LBES8205020
Eyewash bottle, sterilised, diphoterine, Esculape
Eerste-hulpbenodigdheden Oogdouches
Sterilised individual eye wash containing diphoterine.
  • Neutralises and evacuates an acid projection, basic or solvent
  • Sealed ensuring the integrity of the device
  • Disposable
Shelf life: Two years
Leveringsinformatie: Bottle is packed in a storage pouch with belt loop.