Volumetric overflow flasks, without stopper
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0372-10-432 0372-10-365 0372-10-250 0372-10-164 0372-10-094 0372-10-100
NEUB0372-10-432EA 56.4 EUR
NEUB0372-10-432 NEUB0372-10-365 NEUB0372-10-250 NEUB0372-10-164 NEUB0372-10-094 NEUB0372-10-100
Volumetric overflow flasks, without stopper
Kolven Maatkolven
These volumetric overflow flasks are made of borosilicate glass 3.3.
  • Calibrated to deliver (TD, Ex)
  • Cut and ground exactly at the volume-mark
  • Without stopper
Specially designed for water analysis.