Capillary blood collection tubes, MiniCollect®, Vacuette®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
450530_2000 450531_2000 450533_2000 450537_50 450532_2000 450534_2000 450535_2000 450536_2000 450539_2000 450541_2000 450545_2000
GREI450530_2000EA 290 EUR
GREI450530_2000 GREI450531_2000 GREI450533_2000 GREI450537_50 GREI450532_2000 GREI450534_2000 GREI450535_2000 GREI450536_2000 GREI450539_2000 GREI450541_2000 GREI450545_2000
Capillary blood collection tubes, MiniCollect®, Vacuette®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
MiniCollect® offers a gentle way to collect small blood samples for a wide range of analyses for young children, geriatric patients as well as patients with fragile veins.
  • Tube includes an integrated scoop, allowing droplets of blood to be collected easily and hygienically
  • With or without additives
  • The wide opening of the tube ensures an efficient blood flow
  • Minimises adhesion to the wall of the tube and facilitates the mixing of the sample
  • Coagulation tube is intended only for use with venous blood